Learn More
Biomatik Corporation Recombinant Bovine Prolactin (PRL)
Supplier:  Biomatik Corporation RPU55252
Encompass_Preferred
Recombinant Bovine Prolactin (PRL) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Bovine (Bos taurus). Target Name: Prolactin (PRL). Target Synonyms: LTH; Luteotropic Hormone. Accession Number: P01239. Gene ID: 280901. Expression Region: Thr31~Cys229. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0284