Learn More
Biomatik Corporation Recombinant Human Plasmin/Antiplasmin Complex (PAP)
Supplier:  Biomatik Corporation RPU55605
Encompass_Preferred
Recombinant Human Plasmin/Antiplasmin Complex (PAP) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Plasmin/Antiplasmin Complex (PAP). Accession Number: P08697; P00747. Expression Region: Asn40~Lys491+GGGGS*3+Val581~Asn810. Tag Info: N-terminal His Tag. Theoretical MW: 72kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0310