Learn More
Biomatik Corporation Recombinant Pig Interleukin 15 (IL15)
Supplier:  Biomatik Corporation RPU56195
Encompass_Preferred
Recombinant Pig Interleukin 15 (IL15) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Pig (Sus scrofa; Porcine). Target Name: Interleukin 15 (IL15). Accession Number: Q95253. Gene ID: 397683. Expression Region: Thr49~Ser162. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0331