Learn More
Biomatik Corporation Recombinant Moloney murine leukemia virus Gag polyprotein (gag), partial
Supplier:  Biomatik Corporation RPC20279
Encompass_Preferred
Recombinant Moloney murine leukemia virus Gag polyprotein (gag), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Moloney murine leukemia virus (isolate Shinnick) (MoMLV). Target Name: gag. Target Synonyms: Core polyprotein. Accession Number: P03332. Expression Region: 216~478aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 57.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0399