Learn More
Biomatik Corporation Recombinant Macaca mulatta CD40 ligand (CD40LG), partial
Supplier:  Biomatik Corporation RPC21055
Encompass_Preferred
Recombinant Macaca mulatta CD40 ligand (CD40LG), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Macaca mulatta (Rhesus macaque). Target Name: CD40LG. Target Synonyms: Tumor necrosis factor ligand superfamily member 5CD_antigen: CD154. Accession Number: P63304. Expression Region: 44~261aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 28.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0445