Learn More
Biomatik Corporation Recombinant Human Glutathione S Transferase Mu 1 (GSTM1)
Supplier:  Biomatik Corporation RPU41673
Encompass_Preferred
Recombinant Human Glutathione S Transferase Mu 1 (GSTM1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Glutathione S Transferase Mu 1 (GSTM1). Target Synonyms: GST-M1; GST1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GTH4; GTM1; H-B; MU1; GST HB subunit 4. Accession Number: P09488. Gene ID: 2944. Expression Region: Met1~Lys218. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9960