Learn More
Biomatik Corporation Recombinant Mouse Hepcidin (Hepc)
Supplier:  Biomatik Corporation RPU41887
Encompass_Preferred
Recombinant Mouse Hepcidin (Hepc) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Hepcidin (Hepc). Target Synonyms: HAMP; HFE2B; PLTR; LEAP1; Hepcidin Antimicrobial Peptide; Liver-expressed antimicrobial peptide 1; Putative liver tumor regressor. Accession Number: Q9EQ21. Gene ID: 84506. Expression Region: Thr24~Thr83. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9975