Learn More
Biomatik Corporation Recombinant Human Histatin 1 (HTN1)
Supplier:  Biomatik Corporation RPU41917
Encompass_Preferred
Recombinant Human Histatin 1 (HTN1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Histatin 1 (HTN1). Target Synonyms: PPB; HIS1; Histidine-rich protein 1; Post-PB protein. Accession Number: P15515. Gene ID: 3346. Expression Region: Asp20~Asn57. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9978