Learn More
Biomatik Corporation Recombinant Human Interleukin 12 Receptor Beta 2 (IL12Rb2)
Supplier:  Biomatik Corporation RPU42218
Encompass_Preferred
Recombinant Human Interleukin 12 Receptor Beta 2 (IL12Rb2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interleukin 12 Receptor Beta 2 (IL12Rb2). Target Synonyms: IL12R-B2; IL12-RB2; RP11-102M16.1. Accession Number: Q99665. Gene ID: 3595. Expression Region: Tyr559~Gln647. Tag Info: N-terminal His and GST Tag. Theoretical MW: 40kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9989