Learn More
Biomatik Corporation Recombinant Mouse Prostate Specific Antigen (PSA)
Supplier:  Biomatik Corporation RPU42359
Encompass_Preferred
Recombinant Mouse Prostate Specific Antigen (PSA) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Prostate Specific Antigen (PSA). Target Synonyms: KLK3; APS; KLK2A1; HK3; KLKB1; Seminin; Prostate Specific Antigen; Kallikrein-Related Peptidase 3; Semenogelase, g-Seminoprotein; P-30 Antigen; Plasma kallikrein. Accession Number: P00756. Gene ID: 18050. Expression Region: His39~Pro261. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9997