Learn More
Biomatik Corporation Recombinant Mouse Leucine Rich Alpha-2-Glycoprotein 1 (LRG1)
Supplier:  Biomatik Corporation RPU42511
Encompass_Preferred
Recombinant Mouse Leucine Rich Alpha-2-Glycoprotein 1 (LRG1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Leucine Rich Alpha-2-Glycoprotein 1 (LRG1). Accession Number: Q91XL1. Gene ID: 76905. Expression Region: Ser136~Leu342. Tag Info: N-terminal His and GST Tag. Theoretical MW: 53kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0005