Learn More
Biomatik Corporation Recombinant Human Lymphocyte Function Associated Antigen 1 Alpha (CD11a)
Supplier:  Biomatik Corporation RPU42602
Encompass_Preferred
Recombinant Human Lymphocyte Function Associated Antigen 1 Alpha (CD11a) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Lymphocyte Function Associated Antigen 1 Alpha (CD11a). Target Synonyms: LFA1a; ITG-AL; LFA-1; LFA1-A; ITGAL; Integrin,Alpha L; CD11 antigen-like family member A; Leukocyte function-associated molecule 1 alpha chain. Accession Number: P20701. Gene ID: 3683. Expression Region: Asn809~His1066. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0010