Learn More
Biomatik Corporation Recombinant Human Lysozyme (LZM)
Supplier:  Biomatik Corporation RPU42618
Encompass_Preferred
Recombinant Human Lysozyme (LZM) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Lysozyme (LZM). Target Synonyms: LYZ; Renal Amyloidosis; N-Acetylmuramide Glycanhydrolase; Muramidase; 1,4-beta-N-acetylmuramidase C. Accession Number: P61626. Gene ID: 4069. Expression Region: Cys24~Gln144. Tag Info: N-terminal His Tag. Theoretical MW: 14kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0012