Learn More
Biomatik Corporation Recombinant Human Matrix Metalloproteinase 9 (MMP9)
Supplier:  Biomatik Corporation RPU42720
Encompass_Preferred
Recombinant Human Matrix Metalloproteinase 9 (MMP9) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Matrix Metalloproteinase 9 (MMP9). Target Synonyms: GELB; Gelatinase B; CLG4B; CLG4-B; 92 KDa Gelatinase; 92kDa Type IV Collagenase. Accession Number: P14780. Gene ID: 4318. Expression Region: Gly213~Ala399. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0015