Learn More
Biomatik Corporation Recombinant Human Complement Component 3a (C3a)
Supplier:  Biomatik Corporation RPU40948
Encompass_Preferred
Recombinant Human Complement Component 3a (C3a) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Complement Component 3a (C3a). Accession Number: P01024. Gene ID: 718. Expression Region: Ser672~Arg748. Tag Info: N-terminal His and GST Tag. Theoretical MW: 39kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9910