Learn More
Biomatik Corporation Recombinant Mouse N-Acetyl Alpha-D-Glucosaminidase (NAGLU)
Supplier:  Biomatik Corporation RPU42913
Encompass_Preferred
Recombinant Mouse N-Acetyl Alpha-D-Glucosaminidase (NAGLU) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: N-Acetyl Alpha-D-Glucosaminidase (NAGLU). Target Synonyms: NAG; UFHSD1; NAGLU; Sanfilippo Disease IIIB; N-Acetylglucosaminidase,Alpha. Accession Number: O88325. Gene ID: 27419. Expression Region: Arg429~Lys710. Tag Info: N-terminal His and GST Tag. Theoretical MW: 61kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0023