Learn More
Biomatik Corporation Recombinant Human Pentraxin 3, Long (PTX3)
Supplier:  Biomatik Corporation RPU43194
Encompass_Preferred
Recombinant Human Pentraxin 3, Long (PTX3) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Pentraxin 3, Long (PTX3). Target Synonyms: TNFAIP5; TSG-14; Pentaxin-Related Gene,Rapidly Induced By IL-1 Beta; Tumor Necrosis Factor Alpha-Induced Protein 5; Tumor necrosis factor-inducible gene 14 protein. Accession Number: P26022. Gene ID: 5806. Expression Region: Ile183~Ser381. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0032