Learn More
Biomatik Corporation Recombinant Human Phosphatase And Tensin Homolog (PTEN)
Supplier:  Biomatik Corporation RPU43263
Encompass_Preferred
Recombinant Human Phosphatase And Tensin Homolog (PTEN) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Phosphatase And Tensin Homolog (PTEN). Target Synonyms: BZS; MHAM; MMAC1; PTEN1; TEP1; Mutated In Multiple Advanced Cancers 1; Phosphatidylinositol 3,4,5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN. Accession Number: P60484. Gene ID: 5728. Expression Region: Thr2~Val403. Tag Info: N-terminal His Tag. Theoretical MW: 55kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0039