Learn More
Biomatik Corporation Recombinant Human Platelet Factor 4 (PF4)
Supplier:  Biomatik Corporation RPU43341
Encompass_Preferred
Recombinant Human Platelet Factor 4 (PF4) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Platelet Factor 4 (PF4). Target Synonyms: CXCL4; SCYB4; Chemokine C-X-C-Motif Ligand 4; Oncostatin-A; Iroplact. Accession Number: P02776. Gene ID: 5196. Expression Region: Ala31~Ser101. Tag Info: N-terminal His Tag. Theoretical MW: 14kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0042