Learn More
Biomatik Corporation Recombinant Mouse Thrombospondin 1 (THBS1)
Supplier:  Biomatik Corporation RPU43988
Encompass_Preferred
Recombinant Mouse Thrombospondin 1 (THBS1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Thrombospondin 1 (THBS1). Target Synonyms: TSP1; Thrombospondin-1p180. Accession Number: P35441. Expression Region: Ile61~Ile259. Tag Info: N-terminal His Tag. Theoretical MW: 25kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0065