Learn More
Biomatik Corporation Recombinant Human Titin (TTN)
Supplier:  Biomatik Corporation RPU44054
Encompass_Preferred
Recombinant Human Titin (TTN) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Titin (TTN). Target Synonyms: CMD1G; CMH9; CMPD4; HMERF; LGMD2J; TMD; Cardiomyopathy,Dilated 1G(Autosomal Dominant). Accession Number: Q8WZ42. Gene ID: 7273. Expression Region: Pro33779~Gln34025. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0069