Learn More
Biomatik Corporation Recombinant Human Topoisomerase II (TOP2)
Supplier:  Biomatik Corporation RPU44087
Encompass_Preferred
Recombinant Human Topoisomerase II (TOP2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Topoisomerase II (TOP2). Target Synonyms: DNA topoisomerase 2-alpha; DNA topoisomerase II, alpha isozyme. Accession Number: P11388. Gene ID: 7153. Expression Region: Ile923~Arg1148. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0071