Learn More
Biomatik Corporation Recombinant Human Transmembrane Protease, Serine 2 (TMPRSS2)
Supplier:  Biomatik Corporation RPU44149
Encompass_Preferred
Recombinant Human Transmembrane Protease, Serine 2 (TMPRSS2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Transmembrane Protease, Serine 2 (TMPRSS2). Target Synonyms: PRSS10; Serine protease 10. Accession Number: O15393. Gene ID: 7113. Expression Region: Ser284~Gly492. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0075