Learn More
Biomatik Corporation Recombinant Human Troponin I Type 1, Slow Skeletal (TNNI1)
Supplier:  Biomatik Corporation RPU44183
Encompass_Preferred
Recombinant Human Troponin I Type 1, Slow Skeletal (TNNI1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Troponin I Type 1, Slow Skeletal (TNNI1). Target Synonyms: TNN1; Troponin I, slow-twitch isoform. Accession Number: P19237. Gene ID: 7135. Expression Region: Met1~Asp103linkedwithLeu142~Gln187. Tag Info: N-terminal His Tag. Theoretical MW: 24kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0077