Learn More
Biomatik Corporation Recombinant Human Fc Fragment Of IgG Receptor Transporter Alpha (FcgRT)
Supplier:  Biomatik Corporation RPU44400
Encompass_Preferred
Recombinant Human Fc Fragment Of IgG Receptor Transporter Alpha (FcgRT) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Fc Fragment Of IgG Receptor Transporter Alpha (FcgRT). Target Synonyms: FCRN; Neonatal Fc receptor; IgG receptor FcRn large subunit p51. Accession Number: P55899. Gene ID: 2217. Expression Region: Ala24~Ala365. Tag Info: N-terminal His Tag. Theoretical MW: 41kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0085