Learn More
Biomatik Corporation Native Pig Albumin (ALB)
Supplier:  Biomatik Corporation RPU50319
Encompass_Preferred
Native Pig Albumin (ALB) is a purified Natural Protein. Purity: >90% by SDS-PAGE. Host: Pig. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Pig (Sus scrofa; Porcine). Target Name: Albumin (ALB). Target Synonyms: Serum albumin. Accession Number: P08835. Gene ID: 396960. Theoretical MW: 68kDa. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0092