Learn More
Biomatik Corporation Recombinant Human Integrin Alpha V (ITGaV)
Supplier:  Biomatik Corporation RPU51516
Encompass_Preferred
Recombinant Human Integrin Alpha V (ITGaV) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Integrin Alpha V (ITGaV). Target Synonyms: CD51; ITG-AV; MSK8; VNRA; Vitronectin Receptor,Alpha Polypeptide. Accession Number: P06756. Gene ID: 3685. Expression Region: Pro560~Trp744. Tag Info: N-terminal His Tag. Theoretical MW: 24kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0146