Learn More
Biomatik Corporation Recombinant Human R-Spondin 1 (RSPO1)
Supplier:  Biomatik Corporation RPU52361
Encompass_Preferred
Recombinant Human R-Spondin 1 (RSPO1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: R-Spondin 1 (RSPO1). Target Synonyms: RSPONDIN; Roof plate-specific spondin-1. Accession Number: Q2MKA7. Gene ID: 284654. Expression Region: Ser21~Ala135. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0187