Learn More
Biomatik Corporation Recombinant Human Cyclin Dependent Kinase Inhibitor 2A (CDKN2A)
Supplier:  Biomatik Corporation RPU41086
Encompass_Preferred
Recombinant Human Cyclin Dependent Kinase Inhibitor 2A (CDKN2A) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cyclin Dependent Kinase Inhibitor 2A (CDKN2A). Target Synonyms: P16; ARF; CDK4I; CMM2; INK4; INK4a; MLM; MTS1; TP16; P14; P14ARF; P16INK4a; P19; Melanoma P16; Cyclin-dependent kinase 4 inhibitor A; Multiple tumor suppressor 1. Accession Number: P42771. Gene ID: 1029. Expression Region: Ser8~Asp156. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9917