Learn More
Biomatik Corporation Recombinant Rabbit Vascular Endothelial Growth Factor A (VEGFA)
Supplier:  Biomatik Corporation RPU54300
Encompass_Preferred
Recombinant Rabbit Vascular Endothelial Growth Factor A (VEGFA) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rabbit (Oryctolagus cuniculus). Target Name: Vascular Endothelial Growth Factor A (VEGFA). Target Synonyms: VEGF; VEGF-A; VPF; Vascular Endothelial Cell Growth Factor A; Vascular Permeability Factor. Accession Number: Q866G4. Expression Region: Leu1~Lys120. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0240