Learn More
Biomatik Corporation Recombinant Chinese hamster Monocyte Chemotactic Protein 1 (MCP1)
Supplier:  Biomatik Corporation RPU55477
Encompass_Preferred
Recombinant Chinese hamster Monocyte Chemotactic Protein 1 (MCP1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Chinese hamster. Target Name: Monocyte Chemotactic Protein 1 (MCP1). Target Synonyms: CCL2; GDCF2; HC11; HSMCR30; MCAF; MCP1; SCYA2; SMC-CF; Chemokine C-C-Motif Ligand 2; Monocyte Chemotactic And Activating Factor; Monocyte Secretory Protein JE. Accession Number: G3GTT2. Gene ID: 100763833. Expression Region: Gln25~Asn143. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0299