Learn More
Biomatik Corporation Human Caspase 3 (CASP3), Active Protein
Supplier:  Biomatik Corporation RPU56326
Encompass_Preferred
Human Caspase 3 (CASP3), Active Protein is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Caspase 3 (CASP3). Target Synonyms: CPP32; CPP32B; SCA1; Apoptain; Yama; Apoptosis-Related Cysteine Peptidase; Cysteinyl Aspartate Specific Proteinases 3; SREBP cleavage activity 1. Accession Number: p42574. Gene ID: 836. Expression Region: Ser29~His277. Tag Info: N-terminal His Tag. Theoretical MW: 14/18/20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0336