Learn More
Biomatik Corporation Recombinant Mouse Alpha-1-Antitrypsin (a1AT)
Supplier:  Biomatik Corporation RPU40169
Encompass_Preferred
Recombinant Mouse Alpha-1-Antitrypsin (a1AT) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Alpha-1-Antitrypsin (a1AT). Target Synonyms: SERPINA1; SPAAT; A1-AT; PI; Serpin Peptidase Inhibitor,Clade A(Alpha-1 Antiproteinase/AntiTrypsin)Member 1; Alpha-1 protease inhibitor; Short peptide from AAT; Serpin A1. Accession Number: P07758. Gene ID: 20700. Expression Region: Glu25~Lys413. Tag Info: N-terminal His Tag. Theoretical MW: 45kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9871