Learn More
Biomatik Corporation Mouse Platelet Factor 4 (PF4), Active Protein
Supplier:  Biomatik Corporation RPU57826
Encompass_Preferred
Mouse Platelet Factor 4 (PF4), Active Protein is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Platelet Factor 4 (PF4). Target Synonyms: CXCL4; SCYB4; Chemokine C-X-C-Motif Ligand 4; Oncostatin-A; Iroplact. Accession Number: Q9Z126. Gene ID: 56744. Expression Region: Val30~Ser105. Tag Info: N-terminal His Tag. Theoretical MW: 13kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0348