Learn More
Biomatik Corporation Recombinant Human Hyaluronidase-2 (HYAL2)
Supplier:  Biomatik Corporation RPC20266
Encompass_Preferred
Recombinant Human Hyaluronidase-2 (HYAL2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HYAL2. Target Synonyms: Hyaluronoglucosaminidase-2Lung carcinoma protein 2; LuCa-2. Accession Number: Q12891. Expression Region: 21~448aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 51.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0394