Learn More
Biomatik Corporation Recombinant Proteus mirabilis Serralysin (zapA)
Supplier:  Biomatik Corporation RPC20330
Encompass_Preferred
Recombinant Proteus mirabilis Serralysin (zapA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Proteus mirabilis. Target Name: zapA. Target Synonyms: Extracellular domain metalloprotease. Accession Number: Q11137. Expression Region: 17~491aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 68.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0404