Learn More
Biomatik Corporation Recombinant Human Carboxypeptidase A6 (CPA6)
Supplier:  Biomatik Corporation RPC21127
Encompass_Preferred
Recombinant Human Carboxypeptidase A6 (CPA6) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: CPA6. Target Synonyms: CPA6; CPAHCarboxypeptidase A6; EC 3.4.17. Accession Number: Q8N4T0. Expression Region: 130~437aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 52.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0457