Learn More
Biomatik Corporation Recombinant Mouse Fas apoptotic inhibitory molecule 3 (Fcmr), partial
Supplier:  Biomatik Corporation RPC21299
Encompass_Preferred
Recombinant Mouse Fas apoptotic inhibitory molecule 3 (Fcmr), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Faim3. Target Synonyms: Regulator of Fas-induced apoptosis Toso. Accession Number: A1KXC4. Expression Region: 18~262aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 43.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0475