Learn More
Biomatik Corporation Recombinant Human Holliday junction recognition protein (HJURP)
Supplier:  Biomatik Corporation RPC21452
Encompass_Preferred
Recombinant Human Holliday junction recognition protein (HJURP) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HJURP. Target Synonyms: 14-3-3-associated AKT substrateFetal liver-expressing gene 1 proteinUp-regulated in lung cancer 9. Accession Number: Q8NCD3. Expression Region: 1~748aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 99.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0489