Learn More
Biomatik Corporation Recombinant Human Cystatin C (Cys-C)
Supplier:  Biomatik Corporation RPU41105
Encompass_Preferred
Recombinant Human Cystatin C (Cys-C) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cystatin C (Cys-C). Target Synonyms: CST3; Cystatin 3; Gamma Trace; Post-Gamma-Globulin; Neuroendocrine Basic Polypeptide; Amyloid Angiopathy And Cerebral Hemorrhage. Accession Number: P01034. Gene ID: 1471. Expression Region: Ser27~Ala146. Tag Info: N-terminal His Tag. Theoretical MW: 15/33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9921