Learn More
Biomatik Corporation Recombinant Human Ionized Calcium-binding Adapter Molecule 1 (IBA1)
Supplier:  Biomatik Corporation RPU40160
Encompass_Preferred
Recombinant Human Ionized Calcium-binding Adapter Molecule 1 (IBA1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Ionized Calcium-binding Adapter Molecule 1 (IBA1). Target Synonyms: AIF1; IRT1; Daintain; Protein G1; Allograft Inflammatory Factor 1. Accession Number: P55008. Gene ID: 199. Expression Region: Met1~Pro147. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9870