Learn More
Biomatik Corporation Recombinant Human Salivary Alpha Amylase (AMY1A)
Supplier:  Biomatik Corporation RPU40230
Encompass_Preferred
Recombinant Human Salivary Alpha Amylase (AMY1A) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Salivary Alpha Amylase (AMY1A). Target Synonyms: AMY1; AMY1B; AMY1C; Ptyalin; 1,4-alpha-D-glucan glucanohydrolase 1. Accession Number: P04745. Gene ID: 276. Expression Region: Ala15~Leu511. Tag Info: N-terminal His Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9874