Learn More
Biomatik Corporation Recombinant Human Androgen Receptor (AR)
Supplier:  Biomatik Corporation RPU40237
Encompass_Preferred
Recombinant Human Androgen Receptor (AR) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Androgen Receptor (AR). Target Synonyms: AIS; DHTR; HUMARA; KD; NR3C4; SBMA; SMAX1; TFM; Dihydrotestosterone receptor; Nuclear Receptor Subfamily 3,Group C,Member 4; Testicular Feminization; Kennedy Disease. Accession Number: P10275. Expression Region: Pro15~Ser246. Tag Info: N-terminal His Tag. Theoretical MW: 36kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9876