Learn More
Biomatik Corporation Recombinant Human Anterior Gradient 2 (AGR2)
Supplier:  Biomatik Corporation RPU40319
Encompass_Preferred
Recombinant Human Anterior Gradient 2 (AGR2) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Anterior Gradient 2 (AGR2). Target Synonyms: GOB4; HAG2; XAG2; AG2; PDIA17; HPC8; Anterior Gradient Protein 2; Protein Disulfide Isomerase Family A,Member 17; Secreted cement gland protein XAG-2 homolog. Accession Number: O95994. Gene ID: 10551. Expression Region: Arg21~Leu175. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9880