Learn More
Biomatik Corporation Recombinant Human Apolipoprotein H (APOH)
Supplier:  Biomatik Corporation RPU40364
Encompass_Preferred
Recombinant Human Apolipoprotein H (APOH) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Apolipoprotein H (APOH). Target Synonyms: Apo-H; B2G1; BG; B2-GP1; B2GP1; Autoantigen β2 GP I; Previously; Beta 2 Glycoprotein 1; APC inhibitor; Activated protein C-binding protein; Anticardiolipin cofactor; Beta-2-GPI. Accession Number: P02749. Gene ID: 350. Expression Region: Thr22~Cys345. Tag Info: N-terminal His Tag. Theoretical MW: 37kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9883