Learn More
Biomatik Corporation Recombinant Mouse Beta-2-Microglobulin (b2M)
Supplier:  Biomatik Corporation RPU40444
Encompass_Preferred
Recombinant Mouse Beta-2-Microglobulin (b2M) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Beta-2-Microglobulin (b2M). Target Synonyms: BMG; B2MG. Accession Number: P01887. Gene ID: 12010. Expression Region: Gln22~Met119. Tag Info: N-terminal His Tag. Theoretical MW: 14kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9886