Learn More
Biomatik Corporation Recombinant Human Brain Derived Neurotrophic Factor (BDNF)
Supplier:  Biomatik Corporation RPU40514
Encompass_Preferred
Recombinant Human Brain Derived Neurotrophic Factor (BDNF) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Brain Derived Neurotrophic Factor (BDNF). Target Synonyms: Neurotrophin; Abrineurin. Accession Number: P23560. Gene ID: 627. Expression Region: His129~Arg247. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9890