Learn More
Biomatik Corporation Recombinant Human Beta Catenin (β-catenin)
Supplier:  Biomatik Corporation RPU40702
Encompass_Preferred
Recombinant Human Beta Catenin (β-catenin) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Beta Catenin (β-catenin). Target Synonyms: CTNNb1; bCat; b-Cat; CTNN-B1; Catenin Beta 1; Cadherin-Associated Protein. Accession Number: P35222. Gene ID: 1499. Expression Region: Cys439~Ser675. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9896