Learn More
Biomatik Corporation Recombinant Human Caveolin 1 (CAV1)
Supplier:  Biomatik Corporation RPU40740
Encompass_Preferred
Recombinant Human Caveolin 1 (CAV1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Caveolin 1 (CAV1). Target Synonyms: VIP21; Caveolae Protein 22kDa. Accession Number: Q03135. Gene ID: 857. Expression Region: Ser2~Ser104. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9898