Learn More
Biomatik Corporation Recombinant Mouse Enolase 1 (ENO1)
Supplier:  Biomatik Corporation RPU41285
Encompass_Preferred
Recombinant Mouse Enolase 1 (ENO1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Enolase 1 (ENO1). Target Synonyms: NNE; ENO1L1; MPB1; PPH; Alpha Enolase; Enolase 1; Phosphopyruvate hydratase; Plasminogen-binding protein; 2-phospho-D-glycerate hydro-lyase; C-myc promoter-binding. Accession Number: P17182. Gene ID: 13806. Expression Region: Met1~Lys434. Tag Info: N-terminal His Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9934